Hypersil™ Phenyl (Ph) Reversed Phase HPLC Column, 3 μm, 4.6 mm x 150 mm
Product Image
Hypersil™ Phenyl (Ph) Reversed Phase HPLC Column, 3 μm, 4.6 mm x 150 mm
Column Format: Analytical Column; Carbon Load: 5%; Pore Size: 120 Å; Column Phase: Reversed Phase
Have Questions?
Catalog NumberLength (Metric)
HYP30903-154630150 mm
Catalog number HYP30903-154630
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
150 mm
  • For the separation of certain aromatic compounds and moderately polar compounds
  • Retention characteristics similar to Hypersil MOS
  • Alternative selectivity to C8
Specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)150 mm
Particle Size3 μm
pH2 to 8
Pore Size120 Å
Surface Area170 m²/g
USP TypeL11
Stationary PhasePhenyl (Ph)
Packing MaterialPhenyl (Ph)
Carbon Load5%
Column TypeReversed Phase
EndcappedNo
PhaseReversed Phase
Unit SizeEA