Check Order Status
15% Graphite/85% Vespel Ferrules
15% Graphite/85% Vespel Ferrules
Mechanically robust for a long lifetime and are compatible with GC/MS
Have Questions?
Catalog Number
Temperature (Metric) Max
HYP290VT155
350°C
Catalog number
HYP290VT155
Price (MYR)
503.00
PK
Add to cart
Temperature (Metric) Max:
350°C
Price (MYR)
503.00
PK
Add to cart
Form a strong grip with the column and cannot be reused as they form a permanant seal with the column
Temperature limit of 350°C, but must be retightened after initial temperature cycles
Specifications
Description
Ferrules
For Use With (Application)
GC/MS
Temperature (Metric) Max
350°C
For Use With (Equipment)
Thermo Scientific Trace Cold On-Column Injectors
Material
15% Graphite, 85% Vespel