Search Thermo Fisher Scientific
Catalog Number | Material | Diameter (Metric) |
---|---|---|
HYP290VA192 | 15% Graphite, 85% Vespel | 0.32 mm |
290GA139 also known as 072635TH | 100% Graphite | 0.1 to 0.32 mm |
290GA140 | 100% Graphite | 0.45 to 0.53 mm |
290VA191 | 15% Graphite, 85% Vespel | 0.1 to 0.25 mm |
290VA192 | 15% Graphite, 85% Vespel | 0.32 mm |
290VA193 | 15% Graphite, 85% Vespel | 0.45 to 0.53 mm |
Ordering Information
Packaged in contaminant-free, individual blister packs.