Check Order Status
TraceGOLD TG-ALC Plus GC Column
Actual product may vary
TraceGOLD TG-ALC Plus GC Column
Employ the most widely used MS phase in gas chromatography with the 5% phenyl phase Thermo Scientific™ TraceGOLD™ TG-ALC GC Column.
Have Questions?
Change view
Catalog Number
Diameter (Metric) Inner
Film Thickness
HYP26063-3390
0.32 mm
1.8 μm
26063-3390
0.32 mm
1.8 μm
2 Options
Catalog number
HYP26063-3390
Price (MYR)
5,400.00
PK
Add to cart
Diameter (Metric) Inner:
0.32 mm
Film Thickness:
1.8 μm
Price (MYR)
5,400.00
PK
Add to cart
Employ the most widely used MS phase in gas chromatography with the 5% phenyl phase Thermo Scientific™ TraceGOLD™ TG-ALC GC Column.
Low polarity phase, 5% diphenyl/95% dimethyl polysiloxane
Low bleed for excellent signal-to-noise ratio, sensitivity and mass spectral integrity
Exceptional inertness ideal for analysis of active compounds
Specifications
Diameter (Metric) Inner
0.32 mm
Max. Temperature
260°C
Film Thickness
1.8 μm
Length (Metric)
30 mm