Acclaim™ 120 C18 Columns
Acclaim™ 120 C18 Columns
Actual product may vary
Acclaim™ 120 C18 Columns
Use these high-efficiency reversed phase C18 columns for maximum resolution in a wide variety of applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Particle Size
HYP068982100 mm2.1 mm2.2 μm
05914250 mm2.1 mm5 μm
HYP05914250 mm2.1 mm5 μm
5914650 mm4.6 mm5 μm
59147100 mm4.6 mm5 μm
59149250 mm4.6 mm5 μm
76186100 mm3 mm3 μm
68982100 mm2.1 mm2.2 μm
063691150 mm3 mm3 μm
63691150 mm3 mm3 μm
HYP063691150 mm3 mm3 μm
Catalog number HYP068982
Price (MYR)
6,936.00
EA
Add to cart
Length (Metric):
100 mm
Diameter (Metric):
2.1 mm
Particle Size:
2.2 μm
Price (MYR)
6,936.00
EA
Add to cart
Improve the separation efficiency in your reversed-phase applications with Thermo Scientific™ Acclaim™ 120 C18 columns. These columns feature an ultrapure silica substrate with extremely low metal content to minimize tailing effects and deliver symmetrical peak shapes. The unique bonding chemistry yields excellent surface coverage to provide highly predictable separations that are unaffected by secondary interactions. Use these high-efficiency columns for maximum resolution in a wide variety of applications.
High Performance Reversed-Phase Columns for Reproducible Results
  • Very high surface coverage and very low metal content together result in columns with excellent efficiencies
  • Reproducible column-to-column performance
  • Extremely low bleed, fully compatible with MS
  • Stable between pH 2 and 8
  • Available in 2.2, 3, and 5μm particle sizes and 4.6, 3.0, and 2.1mm diameters, with an average pore diameter of 120Å
  • Also available in 2.2μm RSLC format for reliable, fast separations
About the Acclaim Family of HPLC Columns
Achieve the high standards set by modern HPLC and LC/MS methods with the Acclaim family of reversed-phase columns. These silica-based columns with advanced column bonding and endcapping technologies provide complementary selectivity, high column efficiencies, and ultra-high resolution for a wide variety of applications.
  • Broad spectrum of stationary phases, including conventional C8 and C18 phases, polar embedded phases, and specialty phases including wide-pore, are available to meet your specific application needs
  • Performance tested to ensure high quality and reliability for pharmaceutical, biopharmaceutical, environmental, food, and other industrial chromatographic separations
Specifications
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
For Use WithLC/MS
Length (Metric)100 mm
Max. Pressure11600 psi (800 bar)
Particle Size2.2 μm
pH2 to 8
Pore Size120 Å
QuantityEach
Surface Area300 m²/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Product LineAcclaim
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load18%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
TechniqueBio-LC
Green FeaturesResponsibly packaged
Unit SizeEA