DNAPac™ PA100 Oligonucleotide HPLC Columns
DNAPac™ PA100 Oligonucleotide HPLC Columns
Actual product may vary
DNAPac™ PA100 Oligonucleotide HPLC Columns
Features strong anion-exchange resin for high-resolution analysis and purification of synthetic and modified oligonucleotides.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)
HYP043010250 mm4 mm
043010
also known as HC043R010R
250 mm4 mm
Catalog number HYP043010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
250 mm
Diameter (Metric):
4 mm
Feature strong anion-exchange resin for high-resolution analysis and purification of synthetic and modified oligonucleotides with Thermo Scientific™ Dionex™ DNAPac™ PA100 Oligonucleotide Columns.
  • 13μm pellicular, nonporous polymeric resin with bound quaternary amine-functionalized MicroBeads™
  • Capable of n, n-1 resolution for oligonucleotides
  • Resolves oligonucleotides with secondary structures
  • Compatible with solvent, high pH, and high temperatures
  • Analyzes phosphorothioate-based clinical samples
  • Provides easy scale-up
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Diameter (Metric)4 mm
Flow Rate1 mL/min
For Use WithASRS, DX-500 BioLC Chromatography System
Length (Metric)250 mm
Max. Pressure4000 psi (275 bar)
Mobile Phase CompatibilitypH 2 to 12.5, 100% compatible with common organic solvents, Ionic form eluents: chloride, acetate, or bromide in lithium, sodium or ammonium forms
Particle Size13 μm
pH2 to 12.5
Pore SizeNonporous
QuantityEach
Temperature90°C
Stationary PhaseQuaternary Ammonium
Product LineDNAPac
Packing MaterialSpherical, Nonporous Polymer
Column TypeIon Exchange
PhaseIon Exchange
TypeAnalytical Column
TechniqueBio-LC
Green FeaturesResponsibly packaged
Unit SizeEA