MyLab™ Custom Kit - Building DNA With Beads
MyLab™ Custom Kit - Building DNA With Beads
Actual product may vary
MyLab™ Custom Kit - Building DNA With Beads
Are you planning a virtual curriculum or looking for experiments ideal for distance learning? With the MyLab™ Custom Kits, you can easily plan your curriculum. Each kit comes with instructions and materials for 2-3 runs of each experiment.
Have Questions?
Catalog NumberGrade
HGS11036 to 15
Catalog number HGS1103
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Grade:
6 to 15
  • Understand DNA base pairing
  • Model the structure of DNA
  • Explore palindromes, restriction enzymes, DNA synthesis, and genetic engineering
  • Includes instructions, 75 colored beads
Specifications
DescriptionBuilding DNA With Beads
Grade6 to 15
Age12 to 20
For Use With (Application)At Home Education
IncludesInstructions, 75 Colored Beads
Quantity1 Kit
Unit SizeCase