Stainless Steel Sloping Lids
Stainless Steel Sloping Lids
Actual product may vary
Stainless Steel Sloping Lids
Recommended for use above 60°C. Stainless Steel Sloping Lid avoids contamination, reduces evaporation and saves energy.
Have Questions?
Catalog NumberFor Use With (Equipment)
GRALU28SAP18, ST12, ST26
Catalog number GRALU28
Price (MYR)
2,555.00
EA
Add to cart
For Use With (Equipment):
SAP18, ST12, ST26
Price (MYR)
2,555.00
EA
Add to cart
  • For use with Sub Aqua Pro Series Unstirred Water Bath
  • Triangular-shaped stainless steel with plastic handle
  • Choose from the following available sizes:
    • 5L (LU6)
    • 12L (LU14)
    • 18 or 26L (LU28)
    • 34L (LU36)
Specifications
For Use With (Equipment)SAP18, ST12, ST26