Refrigerated immersion coolers consists of a cooling coil connected to a refrigeration unit by a flexible pipe. Extract heat continuously, with the bath control unit controlling temperature.
Have Questions?
Catalog Number
Cooling Capacity
GRAC1GR
350w at 20°C, 110w at 0°C
Catalog number GRAC1GR
Price (MYR)
31,282.00
EA
Add to cart
Cooling Capacity:
350w at 20°C, 110w at 0°C
Price (MYR)
31,282.00
EA
Add to cart
Refrigerated immersion coolers consists of a cooling coil connected to a refrigeration unit by a flexible pipe. Extract heat continuously, with the bath control unit controlling temperature.
C1GR 0°C to 40°C
The cooling coil can be continuously immersed in liquids up to 100°C with the cooler switched off and may be used to cool liquid down from 100°C but it is not designed for continuous operation above 40°C.