Sterile Plastic Inoculation Loops (Volume: 1μL)
Sterile Plastic Inoculation Loops (Volume: 1μL)
Actual product may vary
Sterile Plastic Inoculation Loops (Volume: 1μL)
Designed for quantitative procedures such as sampling, serial dilutions, as well as for bacterial inoculation.
Have Questions?
Catalog NumberQuantityPackaging TypeSterility
GRAC1G800 Pouches of 20Microseal Pouch, EcopackGamma-irradiated
Catalog number GRAC1G
Price (MYR)
-
Quantity:
800 Pouches of 20
Packaging Type:
Microseal Pouch, Ecopack
Sterility:
Gamma-irradiated
  • High Impact Polystyrene construction
  • Full length to avoid contamination
  • Disposable
  • Sterile, aseptically manufactured
  • Medium rigidity
  • Culture loops (Length: 200mm)
  • Smooth edges to avoid tearing and digging in agar
  • Volume: 1μL
  • Color: Blue
Specifications
DisposableSingle-use
Diameter (English) Inside0.04 in.
Diameter (Metric) Inside1 mm
Length (English)7.87 in.
Length (Metric)200 mm
MaterialHigh-impact Polystyrene
SterilityGamma-irradiated
Volume1 μL
ColorBlue
Product TypePlastic Culture Loops
Length (English) Handle7.87 in.
Length (Metric) Handle200 mm
Quantity800 Pouches of 20
Packaging TypeMicroseal Pouch, Ecopack
CalibratedYes
Unit SizeEA