CD45RA Mouse anti-Human, Oligo, Clone: HI100, BD™ AbSeq ™
CD45RA Mouse anti-Human, Oligo, Clone: HI100, BD™ AbSeq ™
Actual product may vary
CD45RA Mouse anti-Human, Oligo, Clone: HI100, BD™ AbSeq ™
Mouse Monoclonal Antibody
Have Questions?
Catalog number FSP940011
Price (MYR)
-
CD45RA Monoclonal antibody specifically detects CD45RA in Human samples. It is validated for Single Cell 3' Sequencing
Specifications
AntigenCD45RA
ApplicationsSingle Cell 3' Sequencing
ClassificationMonoclonal
CloneClone HI100
ConjugateOligo
FormulationAqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene AliasCD45R; PTPRC; LCA; Leukocyte common antigen
Host SpeciesMouse
Quantity25 Tests
Regulatory StatusRUO
Primary or SecondaryPrimary
Target SpeciesHuman
IsotypeIgG2b κ
Unit SizeEach
Contents & Storage
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions.