Hexanes, 99.9%, Extra Dry, mixture of isomers, AcroSeal™
Hexanes, 99.9%, Extra Dry, mixture of isomers, AcroSeal™
Hexanes, 99.9%, Extra Dry, mixture of isomers, AcroSeal™
Hexanes, 99.9%, Extra Dry, mixture of isomers, AcroSeal™
Hexanes, 99.9%, Extra Dry, mixture of isomers, AcroSeal™
Hexanes, 99.9%, Extra Dry, mixture of isomers, AcroSeal™
Thermo Scientific Chemicals
Hexanes, 99.9%, Extra Dry, mixture of isomers, AcroSeal™
CAS: 92112-69-1 | C6H14
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA610370010
Price (MYR)
996.00
EA
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
996.00
EA
Add to cart
Chemical Identifiers
CAS92112-69-1, 110-54-3
Specifications
Chemical Name or MaterialHexanes
CAS92112-69-1
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.
Suspected of damaging fertility.
H
CAS110-54-3