Locust bean gum
Thermo Scientific Chemicals
Locust bean gum
CAS: 9000-40-2
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Catalog number FSA466672500
Price (MYR)
509.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
509.00
EA
Add to cart
Specifications
Chemical Name or MaterialLocust bean gum
CAS9000-40-2
Quantity250 g
EINECS Number232-541-5