Genipin
Thermo Scientific Chemicals
Genipin
CAS: 6902-77-8 | C11H14O5, C11H14O5 | 226.23 g/mol
Have Questions?
Catalog NumberQuantity
FSA4666400101 g
Catalog number FSA466640010
Price (MYR)
1,304.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
1,304.00
EA
Add to cart
Specifications
Chemical Name or MaterialGenipin
CAS6902-77-8
Quantity1 g
Melting Point121.0°C
Health Hazard 1GHS Signal Word: Danger