Betamethasone valerate
Thermo Scientific Chemicals
Betamethasone valerate
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number FSA464930050
Price (MYR)
2,751.00
Each
Quantity:
5 g
Request bulk or custom format
Price (MYR)
2,751.00
Each
Specifications
Chemical Name or Materialβmethasone valerate
CAS2152-44-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement May damage the unborn child.
Suspected of damaging fertility.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3GHS P Statement Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.