Duloxetine hydrochloride
Product Image
Thermo Scientific Chemicals
Duloxetine hydrochloride
333.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA463140010
Price (MYR)
459.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
459.00
EA
Add to cart
Specifications
Molecular Weight (g/mol)333.87
Product TypeDuloxetine hydrochloride
Quantity1 g
Solubility InformationSolubility aq. soln.: .. Other solubilities: soluble in methanol