Niclosamide, 97+%, Thermo Scientific Chemicals
Niclosamide, 97+%, Thermo Scientific Chemicals
Niclosamide, 97+%, Thermo Scientific Chemicals
Niclosamide, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Niclosamide, 97+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number FSA461840250
Price (MYR)
551.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
551.00
EA
Add to cart
Specifications
Chemical Name or MaterialNiclosamide
CAS50-65-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Very toxic to aquatic life.
Health Hazard 3GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.