Ethinylestradiol, 98%, Thermo Scientific Chemicals
Ethinylestradiol, 98%, Thermo Scientific Chemicals
Ethinylestradiol, 98%, Thermo Scientific Chemicals
Ethinylestradiol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethinylestradiol, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA461550050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialEthinylestradiol
CAS57-63-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement Harmful if swallowed. May cause cancer.
Health Hazard 3GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling. Obtain special instructions before use. Wear protective gloves/protective clothing/eye protection/face protection.