Estriol, 97%, Thermo Scientific Chemicals
Estriol, 97%, Thermo Scientific Chemicals
Estriol, 97%, Thermo Scientific Chemicals
Estriol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Estriol, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA460490050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialEstriol
CAS50-27-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Suspected of causing cancer.
May damage fertility or the unborn child.
Health Hazard 3GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.