Ribavirin, CAS# 36791-04-5, is an antiviral agent and a guanosine analog, which stops viral RNA synthesis and mRNA capping. | CAS: 36791-04-5 | C8H12N4O5 | 244.2 g/mol
Have Questions?
Change view
Quantity:
1 g
Catalog number FSA460480010
Price (MYR)
327.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
327.00
EA
Add to cart
Specifications
Chemical Name or MaterialRibavirin
CAS36791-04-5
Health Hazard 1Reproductive toxicity (category 1B)
Health Hazard 2May damage fertility or the unborn child
Ignition Residue0.25% max.
View more
General Description
Ribavirin is a broad-spectrum antiviral nucleoside analog
It inhibits viral genome synthesis
Applications
Ribavirin is an antiviral compound
Its metabolite, ribavirin 5'-phosphate, inhibits inosine monophosphate dehydrogenase (IMP), which is involved in GTP biosynthesis
RUO – Research Use Only
General References:
Keppeke, G.D.; et al. IMP/GTP balance modulates cytoophidium assembly and IMPDH activity. Cell Div. 2018, 15,13:5.
Li, W.; Shen, F.; Weber, G.; Ribavirin and quercetin synergistically downregulate signal transduction and are cytotoxic in human ovarian carcinoma cells. Oncol Res. 1999, 11(5), 243-7.