Daidzin, 98%
Daidzin, 98%
Daidzin, 98%
Thermo Scientific Chemicals
Daidzin, 98%
CAS: 552-66-9 | C21H20O9 | 416.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
Catalog number FSA460421000
Price (MYR)
655.00
EA
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (MYR)
655.00
EA
Add to cart
Specifications
Chemical Name or MaterialDaidzin
CAS552-66-9
Loss on Drying2.5% max.
Infrared SpectrumAuthentic
Assay Percent Range97.5% min. (HPLC)