Methyl palmitate, 99%, analytical standard for GC
Methyl palmitate, 99%, analytical standard for GC
Methyl palmitate, 99%, analytical standard for GC
Methyl palmitate, 99%, analytical standard for GC
Thermo Scientific Chemicals
Methyl palmitate, 99%, analytical standard for GC
CAS: 112-39-0 | C17H34O2 | 270.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number FSA460390050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl palmitate
CAS112-39-0
Melting Point28.0°C
EINECS Number203-966-3
Boiling Point338.0°C