Cefadroxil, 95-105%
Cefadroxil, 95-105%
Cefadroxil, 95-105%
Thermo Scientific Chemicals
Cefadroxil, 95-105%
Have Questions?
Catalog NumberQuantity
FSA4594700505 g
Catalog number FSA459470050
Price (MYR)
2,690.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
2,690.00
EA
Add to cart
Specifications
Chemical Name or MaterialCefadroxil
CAS66592-87-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee