Riluzole, 98%
Riluzole, 98%
Riluzole, 98%
Thermo Scientific Chemicals
Riluzole, 98%
CAS: 1744-22-5 | C8H5F3N2OS | 234.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA459360010
Price (MYR)
687.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
687.00
EA
Add to cart
Specifications
Chemical Name or MaterialRiluzole
CAS1744-22-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Fatal if swallowed.
Health Hazard 3GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Rinse mouth.