Poly(vinyl alcohol), 98% hydrolyzed, approx. M.W. 13,000
Poly(vinyl alcohol), 98% hydrolyzed, approx. M.W. 13,000
Poly(vinyl alcohol), 98% hydrolyzed, approx. M.W. 13,000
Poly(vinyl alcohol), 98% hydrolyzed, approx. M.W. 13,000
Thermo Scientific Chemicals
Poly(vinyl alcohol), 98% hydrolyzed, approx. M.W. 13,000
CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number FSA458245000
Price (MYR)
575.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
575.00
EA
Add to cart
Specifications
Chemical Name or MaterialPoly(vinyl alcohol)
CAS9002-89-5
Melting Point230.0°C to 240.0°C
Molecular Formula(C2H4O)n
Linear Formula[-CH2CH(OH)-]n