Donepezil hydrochloride
Donepezil hydrochloride
Donepezil hydrochloride
Donepezil hydrochloride
Thermo Scientific Chemicals
Donepezil hydrochloride
CAS: 120011-70-3 | C24H30ClNO3 | 415.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number FSA458050050
Price (MYR)
1,868.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,868.00
EA
Add to cart
Specifications
Chemical Name or MaterialDonepezil hydrochloride
CAS120011-70-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
If eye irritation persists: Get medical advice/