Montelukast sodium, 98%
Montelukast sodium, 98%
Montelukast sodium, 98%
Montelukast sodium, 98%
Thermo Scientific Chemicals
Montelukast sodium, 98%
CAS: 151767-02-1 | C35H35ClNNaO3S | 608.17 g/mol
Have Questions?
Catalog NumberQuantity
FSA4564400101 g
Catalog number FSA456440010
Price (MYR)
1,400.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
1,400.00
EA
Add to cart
Specifications
Chemical Name or MaterialMontelukast sodium
CAS151767-02-1
Health Hazard 1Danger
Health Hazard 2GHS H Statement Causes serious eye damage.
Health Hazard 3GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.