Oleanolic acid, 97%
Oleanolic acid, 97%
Oleanolic acid, 97%
Thermo Scientific Chemicals
Oleanolic acid, 97%
CAS: 508-02-1 | C30H48O3 | 456.7 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA456120050
Price (MYR)
635.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
635.00
EA
Add to cart
Specifications
Chemical Name or MaterialOleanolic acid
CAS508-02-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.