Ketoconazole, 98%
Ketoconazole, 98%
Ketoconazole, 98%
Ketoconazole, 98%
Ketoconazole, CAS # 65277-42-1, is a synthetic, broad-spectrum antifungal agent with the ability to inhibit a variety of fungal infections. | CAS: 65277-42-1 | C26H28Cl2N4O4 | 531.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
Catalog number FSA455470250
Price (MYR)
3,829.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
3,829.00
EA
Add to cart
Specifications
Chemical Name or MaterialKetoconazole
CAS65277-42-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May damage fertility.
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure if swallowed.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
Do not get in eyes,on skin,or on clothing.
Do not eat,drink or smoke
View more
General Description
  • Ketoconazole is an imidazole antifungal agent
  • It functions by inhibiting synthesis of ergosterol, which is the fungal equivalent of cholesterol
Applications
  • Ketoconazole shows fungicide activity against a variety of yeast infections
  • It is an inhibitor of CYP3A4, which is necessary for the conversion of lanosterol to ergosterol
  • It increases fungal cellular permeability
  • It also inhibits endogenous respiration by disrupting membrane phospholipids
  • Ketoconazole can also inhibit the synthesis of thromboxane and sterols such as aldosterone, cortisol, and testosterone
RUO – Research Use Only
General References:
  1. Takano, M.; Hasegawa, R.; Fukuda, T.; Yumoto, R.; Nagai, J.; Murakami, T. Interaction with P-glycoprotein and transport of erythromycin, midazolam and ketoconazole in Caco-2 cells. European Journal of Pharmacology.1998, 358 (3), 289-294.
  2. Goeders, N. E.; Peltier, R. L.; Guerin, G. F. Ketoconazole reduces low dose cocaine self-administration in rats. Drug and Alcohol Dependence.1998, 53 (1), 67-77.