Colistin sulfate salt
Colistin sulfate salt
Colistin sulfate salt
Colistin sulfate salt
Colistin sulfate salt
Colistin Sulfate salt (2:5), CAS # 1264-72-8, (synonym: Polymyxin E Complex), is a complex mixture of antibiotics, with Colistin A and B being the primary components. It has anti-bacterial activity against Gram-positive and Gram-negative bacteria and may also be used as a neuromuscular blocker.
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number FSA455392500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialColistin sulfate salt
TypeColistin Sulfate salt
CAS1264-72-8
Health Hazard 1Danger
Health Hazard 2GHS H Statement Toxic if swallowed.
View more
General description
• Colistin Sulfate salt is a stable, cationic, water-soluble, white powder composed of over 30 components
• Originally isolated from B. polymyxa, it inhibits the growth of the Gram-negative (e.g. E. coli, P. aeruginosa, P. fluorescens, and S. enterica), and Gram-positive bacteria (e.g. L. lactis, P. polymyxa, P. acidilactici, and S. aureus) by permeabilizing the bacterial cell membrane
Applications
• Colistin Sulfate has potent anti-endotoxin activity rendered through its ability to bind lippopolysaccharides that make up bacterial endotoxins
• This compound is often used in the management of multi-drug resistant Gram-negative infections
RUO – Research Use Only
General References:
  1. Falagas, M. E.; Kasiakou, S. K.; Saravolatz, L. D. Colistin: The Revival of Polymyxins for the Management of Multidrug-Resistant Gram-Negative Bacterial Infections Clinical Infectious Diseases. 2005, 40 (9), 1333–1341.
  2. Suzuki, T.; Inouye, H.; Fujikawa, K.; et al. Studies on the chemical structure of colistin. I. Fractionation, molecular weight determination, amino acid and fatty acid composition. J. Biochem. 1963, 54(1), 25-33.
  3. Naghmouchi, K.; Hammami, R.; Fliss, I.; et al. Colistin A and colistin B among inhibitory substances of Paenibacillus polymyxa JB05-01-1. Arch. Microbiol. 2012, 194(5), 363-370.
  4. Mitsugui, C.S.; Tognim, M.C.B.; Cardoso, C.L.; et al. In vitro activity of polymyxins in combination with β-lactams against clinical strains of Pseudomonas aeruginosa. Int. J. Antimicrob. 2011, 38(5), 447-450.