3'-Azido-3'-deoxythymidine, 98%
3'-Azido-3'-deoxythymidine, 98%
3'-Azido-3'-deoxythymidine, 98%
3'-Azido-3'-deoxythymidine, 98%
3'-Azido-3'-deoxythymidine, 98%
CAS: 30516-87-1 | C10H13N5O4 | 267.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA454980010
Price (MYR)
401.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
401.00
EA
Add to cart
Specifications
Chemical Name or Material3'-Azido-3'-deoxythymidine
CAS30516-87-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement Suspected of causing cancer. May cause cancer by inhalation. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Health Hazard 3GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection.