Cefotaxime sodium salt, 95%
Cefotaxime sodium salt, 95%
Cefotaxime sodium salt, 95%
Cefotaxime sodium salt, 95%
Cefotaxime sodium salt, 95%
Cefotaxime sodium salt, CAS # 64485-93-4, is the sodium salt form of cefotaxime with β-lactamase resistant antibiotic activity.
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass Bottle
Catalog number FSA454950250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCefotaxime sodium salt
TypeCefotaxime Sodium Salt
CAS64485-93-4
Melting Point201.0°C to 210.0°C
PackagingGlass Bottle
View more
General Information
• The cefotaxime sodium salt is a - lactamase considered a third-generation cephalosporin with antibiotic and bactericidal activities
• This compound exerts its biological activity by binding to penicillin-binding proteins (PBP) located on the inner membrane of the bacterial cell wall leading to their inactivation. The inactivation of these proteins affects the cross-linking of peptidoglycan chains required for bacterial cell wall strength and rigidity, resulting in the weakening of cell walls and cell lysis.
Applications
• It is an efficient antibacterial compound that acts against gram-negative bacteria, without activity against gram-positive bacteria such as pseudomonas and penicillin-resistant strains of streptococcus pneumonia
• It has been used for plant cell culture and human cell lines in vitro
• Cefotaxime sodium salt has been used in antibiotic susceptibility testing (AST) methods
• Cefotaxime sodium salt is also used to remove bacteria from Ostreococcus tauri cultures
RUO – Research Use Only
General References:
  1. Ali, M. M.; Gaballa, A. S.; Teleb, S. M. Spectroscopic and thermal investigations of charge-transfer complexes formed between cefotaxime sodium drug and various acceptors. Russ. J. Gen. Chem. 2015, 85 (3), 731-745.
  2. Yin, Y.; Bao, Y.; Gao, Z.; Wang, Z.; Liu, D.; Hao, H.; Wang, Y. Solubility of Cefotaxime Sodium in Ethanol + Water Mixtures under Acetic Acid Conditions. J. Chem. Eng. Data 2014, 59 (6), 1865-1871.
  3. Carmine, A.; Brogden, R.; Heel, R.; Speight, T.; Avery, G. Cefotaxime. A review of its antibacterial activity, pharmacological properties and therapeutic use. Drugs. 1983, 25 (3), 223-289.