L-Phenylalanine, 98.5-101.0%, specified according to the requirements of USP
CAS: 63-91-2 | C9H11NO2 | 165.19 g/mol
Have Questions?
Change view
Quantity:
100 g
25 g
Catalog number FSA453931000
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialL-Phenylalanine, 98.5-101.0%
Melting Point270.0°C to 275.0°C
CAS63-91-2
pH5.4 to 6.0 (1% solution)
Chloride0.05% max.