Itraconazole, 99%
Itraconazole, 99%
Itraconazole, 99%
Itraconazole, 99%
Itraconazole, or voriconazole, CAS # 84625-61-6, is an N-arylpiperazine that has antifungal properties. | CAS: 84625-61-6 | C35H38Cl2N8O4 | 705.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number FSA452870250
Price (MYR)
1,922.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,922.00
EA
Add to cart
Specifications
Chemical Name or MaterialItraconazole
CAS84625-61-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
View more
General Description
• Itraconazole is an aromatic ether that works as a strong inhibitor of P-glycoproteins and CYP3A4
• Itraconazole also acts as a hedgehog signaling inhibitor and P450 inhibitor
Applications
• Itraconazole can be exhibits antifungal activity against organisms including aspergillosis, blastomycosis, candidiasis, chromoblastomycosis, coccidioidomycosis, cryptococcosis, histoplasmosis, and sporotrichosis
• Itraconazole may be used to experimentally target the delivery of other molecules, such as plasmids, to cells
RUO – Research Use Only
General References:
  1. Sharma, P.; Bhalla, M.; Thami, G.P.; Chander, J. Evaluation of efficacy and safety of oral terbinafine and itraconazole combination therapy in the management of dermatophytosis. J Dermatolog Treat. 2020, 31(7), 749-753.
  2. Hamed Khater, M.; Khattab, F.M. Combined long-pulsed Nd-Yag laser and itraconazole versus itraconazole alone in the treatment of onychomycosis nails. J Dermatolog Treat. 2020, 31(4), 406-409.