Perchloric acid, extra pure, ca. 70% solution in water
Perchloric acid, extra pure, ca. 70% solution in water
Perchloric acid, extra pure, ca. 70% solution in water
Perchloric acid, extra pure, ca. 70% solution in water
Perchloric acid, extra pure, ca. 70% solution in water
CAS: 7601-90-3 | ClHO4 | 100.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number FSA451770025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Request bulk or custom format
Specifications
Chemical Name or MaterialPerchloric Acid
CAS7601-90-3
Melting Point-18.0°C
Boiling Point203.0°C
Density1.6600g/mL