Sodium hydroxide, Thermo Scientific Chemicals
Sodium hydroxide, Thermo Scientific Chemicals
Sodium hydroxide, Thermo Scientific Chemicals
Sodium hydroxide, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium hydroxide, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
Packaging:
Plastic Bottle
Catalog number FSA451130050
Price (MYR)
522.00
EA
Add to cart
Quantity:
5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
522.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium Hydroxide
Boiling Point1390.0°C
CAS1310-73-2
Melting Point318.0°C
Molecular FormulaHNaO