Ethyl maltol, 98%
Ethyl maltol, 98%
Ethyl maltol, 98%
Thermo Scientific Chemicals
Ethyl maltol, 98%
CAS: 4940-11-8 | C7H8O3 | 140.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number FSA450800250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialEthyl maltol
CAS4940-11-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Health Hazard 3GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.