Ammonium bromide, 98%, extra pure
Ammonium bromide, 98%, extra pure
Ammonium bromide, 98%, extra pure
Ammonium bromide, 98%, extra pure
Thermo Scientific Chemicals
Ammonium bromide, 98%, extra pure
CAS: 12124-97-9 | BrH4N | 97.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic bottle
Catalog number FSA450650010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Boiling Point235.0°C
Chemical Name or MaterialAmmonium bromide
Melting Point452.0°C
CAS12124-97-9
Health Hazard 1GHS Signal Word: Warning