L(+)-Ascorbyl palmitate, specified according to requirements of USP/Ph.Eur., Thermo Scientific Chemicals
L(+)-Ascorbyl palmitate, specified according to requirements of USP/Ph.Eur., Thermo Scientific Chemicals
L(+)-Ascorbyl palmitate, specified according to requirements of USP/Ph.Eur., Thermo Scientific Chemicals
L(+)-Ascorbyl palmitate, specified according to requirements of USP/Ph.Eur., Thermo Scientific Chemicals
Thermo Scientific Chemicals
L(+)-Ascorbyl palmitate, specified according to requirements of USP/Ph.Eur., Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number FSA449481000
Price (MYR)
1,014.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
1,014.00
EA
Add to cart
Specifications
Chemical Name or MaterialL(+)-Ascorbyl palmitate
CAS137-66-6
Melting Point111°C to 115°C
EINECS Number205-305-4
Boiling Point250°C