Prednisolone, 99%, Thermo Scientific Chemicals
Prednisolone, 99%, Thermo Scientific Chemicals
Prednisolone, 99%, Thermo Scientific Chemicals
Prednisolone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Prednisolone, 99%, Thermo Scientific Chemicals
Prednisolone, or metacortandralone, CAS # 50-24-8, is a synthetic glucocorticoid.
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
View all
Glass Bottle
Catalog number FSA449470250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPrednisolone
CAS50-24-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May damage the unborn child.
Health Hazard 3GHS P Statement
Use personal protective equipment as required.
Do not handle until all safety precautions have been read and understood.
View more
General Description
• Prednisolone is a derivative of prednisone with an oxo to beta-hydroxy reduction at position 11
• Prednisolone binds cell surface receptors and is transported to the nucleus, where it binds certain nuclear receptors responsible for transcription of proinflammatory cytokines
Applications
• Prednisolone can be used for laboratory research on tumor growth and prevention
• Prednisolone may also be used for developmental and endocrine research, in vivo or in vitro
RUO – Research Use Only
General References:
  1. Waldrop, M.A.; Karingada, C.; Storey, M.A.; Powers, B.; Lammarino, M.A.; Miller, N.F.; Alfano, L.N.; Noritz, G.; Rossman, I.; Ginsberg, M.; Mosher, K.A., Broomall, E.; Goldstein, J.; Bass, N.; Lowes, L.P.; Tsao, C.Y.; Mendell, J.R.; Connolly, A.M. Gene Therapy for Spinal Muscular Atrophy: Safety and Early Outcomes. Pediatrics. 2020, 146(3), e20200729.
  2. Koumpa, F.S.; Forde, C.T.; Manjaly, J.G. Sudden irreversible hearing loss post COVID-19. BMJ Case Rep. 2020, 13(11), e238419.