Toluene, 99.85%, Extra Dry, AcroSeal™, package of 4x25ML bottles
Toluene, 99.85%, Extra Dry, AcroSeal™, package of 4x25ML bottles
Toluene, 99.85%, Extra Dry, AcroSeal™, package of 4x25ML bottles
Toluene, 99.85%, Extra Dry, AcroSeal™, package of 4x25ML bottles
Thermo Scientific Chemicals
Toluene, 99.85%, Extra Dry, AcroSeal™, package of 4x25ML bottles
CAS: 108-88-3 | C7H8 | 92.14 g/mol
Have Questions?
Catalog NumberQuantity
FSA448401000100 mL
Catalog number FSA448401000
Price (MYR)
329.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
329.00
EA
Add to cart
Specifications
Chemical Name or MaterialToluene
Melting Point-95.0°C
Name NoteExtra Dry
CAS108-88-3