Ammonium iodide, 99%, extra pure
Ammonium iodide, 99%, extra pure
Ammonium iodide, 99%, extra pure
Ammonium iodide, 99%, extra pure
Thermo Scientific Chemicals
Ammonium iodide, 99%, extra pure
Ammonium iodide, 99%, H4IN, CAS Number-12027-06-4, unii-oz8f027ldh, ammonium iodide, hsdb 476, ammoniumiodid, ammonium iodide nh4 i, nh4i, oz8f027ldh, ammonium jodatum, azanium iodide, ammonium-iodid, 500g, 98.5% min. (Argentometry), CHEBI:63040, 2.5100g/mL, 234-717-7, 144.94, Extra Pure | CAS: 12027-06-4 | H4IN | 144.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number FSA448075000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium iodide
Melting Point551.0°C
CAS12027-06-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement