Sodium thiosulfate pentahydrate, 99+%, for biochemistry
Sodium thiosulfate pentahydrate, 99+%, for biochemistry
Sodium thiosulfate pentahydrate, 99+%, for biochemistry
Sodium thiosulfate pentahydrate, 99+%, for biochemistry
Thermo Scientific Chemicals
Sodium thiosulfate pentahydrate, 99+%, for biochemistry
CAS: 10102-17-7 | Na2O3S2·5H2O | 248.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Catalog number FSA447942500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium thiosulfate pentahydrate
Melting Point48.5°C
CAS10102-17-7
Quantity250 g
Assay Percent Range99.0 to 101% (Iodimetry)