Citric acid, monosodium salt, 99%, for analysis, anhydrous
Citric acid, monosodium salt, 99%, for analysis, anhydrous
Citric acid, monosodium salt, 99%, for analysis, anhydrous
Citric acid, monosodium salt, 99%, for analysis, anhydrous
Thermo Scientific Chemicals
Citric acid, monosodium salt, 99%, for analysis, anhydrous
CAS: 18996-35-5 | C6H7NaO7 | 214.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA447692500
Price (MYR)
284.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
284.00
EA
Add to cart
Specifications
Chemical Name or MaterialCitric acid, monosodium salt
CAS18996-35-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Cont
View more
Biological buffer
RUO – Research Use Only