1,2-Propanediol, ACS reagent
1,2-Propanediol, ACS reagent
1,2-Propanediol, ACS reagent
1,2-Propanediol, ACS reagent
Thermo Scientific Chemicals
1,2-Propanediol, ACS reagent
CAS: 57-55-6 | C3H8O2 | 76.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Catalog number FSA447415000
Price (MYR)
444.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
444.00
EA
Add to cart
Specifications
Chemical Name or Material1,2-Propanediol
CAS57-55-6
Melting Point-60.0°C
Density1.0300g/mL
EINECS Number200-338-