Ammonium dihydrogen phosphate, 99%, for HPLC
Ammonium dihydrogen phosphate, 99%, for HPLC
Ammonium dihydrogen phosphate, 99%, for HPLC
Ammonium dihydrogen phosphate, 99%, for HPLC
Thermo Scientific Chemicals
Ammonium dihydrogen phosphate, 99%, for HPLC
CAS: 7722-76-1 | H6NO4P | 115.03 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA447382500250 gPlastic bottle
Catalog number FSA447382500
Price (MYR)
1,547.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,547.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmmonium dihydrogen phosphate
Melting Point190.0°C
CAS7722-76-1
Quantity250 g
EINECS Number231-764-5
View more
For HPLC
RUO – Research Use Only