Acetic acid, ammonium salt, 98%, for biochemistry
Acetic acid, ammonium salt, 98%, for biochemistry
Acetic acid, ammonium salt, 98%, for biochemistry
Acetic acid, ammonium salt, 98%, for biochemistry
Thermo Scientific Chemicals
Acetic acid, ammonium salt, 98%, for biochemistry
CAS: 631-61-8 | C2H7NO2 | 77.083 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA447121000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Absorbance(c=1M, water), (c=1M, water) at 0nm, 0.01 max. at 280nm, 0.01 max. at 350nm, 0.02 max. at 254nm
Chemical Name or MaterialAcetic acid, ammonium salt
CAS631-61-8
Melting Point114.0°C
Assay Percent Range98%
View more
For biochemistry
RUO – Research Use Only