D(+)-Glucose 6-phosphate sodium salt, 98%
D(+)-Glucose 6-phosphate sodium salt, 98%
D(+)-Glucose 6-phosphate sodium salt, 98%
D(+)-Glucose 6-phosphate sodium salt, 98%
Thermo Scientific Chemicals
D(+)-Glucose 6-phosphate sodium salt, 98%
CAS: 54010-71-8 | C6H12NaO9P | 282.12 g/mol
Have Questions?
Catalog NumberQuantity
FSA4469800101 g
Catalog number FSA446980010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialD(+)-Glucose 6-phosphate sodium salt
CAS54010-71-8
Melting Point192°C to 197°C
EINECS Number258-921-
Assay Percent Range98%