2',7'-Dichlorofluorescein, ACS reagent
2',7'-Dichlorofluorescein, ACS reagent
2',7'-Dichlorofluorescein, ACS reagent
2',7'-Dichlorofluorescein, ACS reagent
Thermo Scientific Chemicals
2',7'-Dichlorofluorescein, ACS reagent
CAS: 76-54-0 | C20H10Cl2O5 | 401.195 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Packaging:
Glass bottle
Catalog number FSA446930100
Price (MYR)
951.00
EA
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
951.00
EA
Add to cart
Specifications
Chemical Name or Material2', 7'-Dichlorofluorescein
CAS76-54-0
Molecular FormulaC20H10Cl2O5
Synonym2',7'-Dichloro-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one
InChI KeyVFNKZQNIXUFLBC-UHFFFAOYSA-N