Acetic acid, sodium salt trihydrate, 99+%, for biochemistry
CAS: 6131-90-4 | C2H3NaO2·3H2O | 136.08 g/mol
Have Questions?
Change view
Quantity:
250 g
Packaging:
Plastic bottle
Catalog number FSA446792500
Price (MYR)
250.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
250.00
EA
Add to cart
Specifications
Chemical Name or MaterialAcetic acid, sodium salt trihydrate
CAS6131-90-4
Absorbance0.05 max. (3M) at 260nm, 0.05 max. (3M) at 280nm
Melting Point58.0°C
Assay Percent Range99+%